Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J4649

Sigma-Aldrich

JNJ-10198409

≥98% (HPLC), solid

Synonym(s):

3-Fluoro-N-(6,7-dimethoxy-2,4-dihydroindeno[1,2-c]pyrazol-3-yl)phenylamine, N-(3-fluorophenyl)-2,4-dihydro-6,7-dimethoxy-Indeno[1,2-c]pyrazol-3-amine, RWJ 540973

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$264.00

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$264.00

About This Item

Empirical Formula (Hill Notation):
C18H16FN3O2
CAS Number:
Molecular Weight:
325.34
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL
H2O: <2 mg/mL

originator

Johnson & Johnson

storage temp.

−20°C

SMILES string

COc1cc2Cc3c(Nc4cccc(F)c4)n[nH]c3-c2cc1OC

InChI

1S/C18H16FN3O2/c1-23-15-7-10-6-14-17(13(10)9-16(15)24-2)21-22-18(14)20-12-5-3-4-11(19)8-12/h3-5,7-9H,6H2,1-2H3,(H2,20,21,22)

InChI key

ZDNURMVOKAERHZ-UHFFFAOYSA-N

Biochem/physiol Actions

JNJ-10198409 is a potent ATP-competitive inhibitor of Platelet-Derived Growth Factor receptor tyrosine kinase (PDGF-RTK) with both antiangiogenic and a direct tumor cell antiproliferative activity. It is selective for PDGF-β kinase with IC50 values of 4.2 nM for PDGF-β and 45 nM for PDGF-α kinase.
JNJ-10198409 is an ATP-competitive Platelet-Derived Growth Factor Receptor-beta (PDGFR-β) inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the PAF Receptor and PDGFR pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael R D'Andrea et al.
Molecular cancer therapeutics, 4(8), 1198-1204 (2005-08-12)
With the advent of agents directed against specific molecular targets in drug discovery, it has become imperative to show a compound's cellular impact on the intended biomolecule in vivo. The objective of the present study was to determine if we
Chih Y Ho et al.
Journal of medicinal chemistry, 48(26), 8163-8173 (2005-12-22)
A series of (6,7-dimethoxy-2,4-dihydroindeno[1,2-c]pyrazol-3-yl)phenylamines has been optimized to preserve both potent kinase inhibition activity against the angiogenesis target, the receptor tyrosine kinase of Platelet-Derived Growth Factor-BB (PDGF-BB), and to improve the broad tumor cell antiproliferative activity of these compounds. This

Articles

We offers many products related to PDGFR for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service