Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J4500

Sigma-Aldrich

Anti-c-Jun N-Terminal Kinase antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-JNK1, JNK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen, JNK1 46 kDa
antigen, JNK2 55 kDa

contains

15 mM sodium azide

species reactivity

rat, human, mouse

technique(s)

microarray: suitable
western blot: 1:16,000 using rat brain extract and mouse NIH-3T3 fibroblast extract, respectively
western blot: 1:2,000 using mouse NIH 3T3 fibroblast cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK8(5599)
mouse ... Mapk8(26419)
rat ... Mapk8(116554)

General description

Anti-c-Jun N-Terminal Kinase (JNK1, JNK2) is developed in rabbit using a synthetic peptide, corresponding to amino acids of human c-Jun N-terminal kinase 1 (JNK1), coupled to KLH as the immunogen.
JNK (c-Jun N terminal protein kinase, also known as stress activated protein kinase, SAPK1) is a protein that belongs to serine/threonine-protein kinase family. It has a critical role in inhibiting WOX1 (WW domain-containing oxidoreductase) mediated apoptosis. It also mediates starvation-induced BCL2 phosphorylation and activates autophagy. Anti-c-Jun N-terminal kinase antibody can be used in microarray. Rabbit anti-c-Jun N-terminal kinase antibody reacts specifically with JNK1 (46 kD) and JNK2 (55 kD) of cell culture lysates and rat brain tissue extracts. The product has also shown weak cross reactivity for JNK2β (50 kD) isoform in mouse NIH-3T3 fibroblast cell lysate.

Immunogen

Synthetic peptide corresponding to amino acids 339-354 of human JNK1 (c-Jun N-terminal kinase 1), conjugated to KLH. The sequence is highly conserved in JNK1, JNK2 α, β, γ (p54 SAPK α, β, γ) and identical in human, rat, and mouse JNK1.

Application

Anti-c-Jun N-Terminal Kinase antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Mitogen-activated protein kinases (MAPKs) are serine/threonine kinases which play a central role in mitogenic signaling. The MAPKs function to transduce extracellular signals to intracellular targets, including transcription factors controlling the expression of genes essential to many cellular processes including proliferation, development and differentiation. JNK1 is essential for tumor necrosis factor alpha (TNF-α)-induced c-Jun kinase activation, c-Jun expression, and apoptosis. JNK1 and JNK2 participates in the survival of neuronal cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

c-Jun N-terminal protein kinase 1 (JNK1), but not JNK2, is essential for tumor necrosis factor alpha-induced c-Jun kinase activation and apoptosis
Liu J, et al.
Molecular and Cellular Biology, 24(24), 10844-10856 (2004)
MEK7-dependent activation of p38 MAP kinase in keratinocytes
Dashti SR, et al.
The Journal of Biological Chemistry, 276(11), 8059-8063 (2001)
Induction of COX-2 enzyme and down-regulation of COX-1 expression by lipopolysaccharide (LPS) control prostaglandin E2 production in astrocytes
Font-Nieves M, et al.
The Journal of Biological Chemistry, 287(9), 6454-6468 (2012)
Shao Li et al.
Scientific reports, 6, 39320-39320 (2016-12-23)
Amyloid precursor protein (APP), commonly associated with Alzheimer's disease, also marks axonal degeneration. In the recent studies, we demonstrated that APP aggregated at nodes of Ranvier (NORs) in myelinated central nervous system (CNS) axons and interacted with Nav1.6. However, the
Mitogen-Activated Protein Kinases and Their Role in Radiation Response.
Anupama M and Rajagopal R
Genes & Cancer, 4(9-10), 401-408 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service