Skip to Content
MilliporeSigma
All Photos(1)

Documents

I9905

Sigma-Aldrich

Interleukin-2/Fc Chimera (non-lytic) from mouse

>98% (SDS-PAGE), recombinant, expressed in CHO cells, solution

Synonym(s):

IL-2:Fc (non-lytic)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in CHO cells

Assay

>98% (SDS-PAGE)

form

solution

mol wt

94 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.1 mg/mL

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il2(16183)

Biochem/physiol Actions

IL-2 (interleukin-2) is a cytokine which is involved in the differentiation of T cell and natural killer (NK) cell, T cell memory, activation-induced lymphocyte cell death. The protein also has a role in regulatory T cell (Treg) development and homeostasis. It is linked with the risk of Grave′s disease, rheumatoid arthritis, celiac disease, multiple sclerosis, psoriasis, Crohn′s disease, ulcerative colitis and type-1 diabetes (T1D).
Possesses the biological functions of the IL-2 moiety and a prolonged circulating half-life determined by the Fc domain.

Other Notes

Dimeric fusion protein consisting of mouse IL-2 fused to mutant mouse IgG2a Fc.

Physical form

Solution in 0.2 μm filtered phosphate buffered saline, pH 7.4.

Analysis Note

The biological activity is measured in a cell proliferation assay using CTLL2 cells.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Robb et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(20), 6486-6490 (1984-10-01)
Human interleukin 2 was separated into multiple molecular forms by selective immunoaffinity chromatography and chromatofocusing. For the most part, this heterogeneity was attributed to variations in glycosylation of the threonine residue in position 3 of the polypeptide chain. The various
Parul Mehra et al.
Molecular and cellular biology, 35(22), 3880-3891 (2015-09-10)
Interleukin-2 (IL-2) is a potent cytokine with roles in both immunity and tolerance. Genetic studies in humans and mice demonstrate a role for Il2 in autoimmune disease susceptibility, and for decades the proximal Il2 upstream regulatory region has served as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service