Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9512

Sigma-Aldrich

Indole-3-acetylglycine

suitable for plant cell culture

Synonym(s):

IAA-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

technique(s)

cell culture | plant: suitable

storage temp.

−20°C

SMILES string

OC(=O)CNC(=O)Cc1c[nH]c2ccccc12

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service