Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9401

Sigma-Aldrich

Interleukin-1β human

IL-1β, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-1β, Lymphocyte activating factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤12 pg/mL ED50

quality

endotoxin tested

mol wt

17.3 kDa

packaging

pkg of 5 and 25 μg
pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

color

white to off-white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL1B(3553)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Interleukin-1 (IL-1), originally known as Lymphocyte Activating Factor (LAF), activates T cell lymphocytes, which then proliferate and secrete interleukin-2 (IL-2). IL-1 is released primarily from stimulated macrophages and monocytes, but has also been shown to be released from several other types and is thought to play a key role in inflammatory and immune responses. Other synonyms for IL-1 include: endogenous pyrogen (EP), mitogenic protein (MP), Helper Peak-1 (HP-1), T Cell Replacing Factor III (TRF III or TRFM), B Cell Activating Factor (BAF) and B Cell Differentiation Factor (BDF) The two closely related agents Interleukin-1α (IL-1α) and Interleukin-1β (IL-1β) share 62% homology in amino acid sequence and elicit nearly identical biological responses. IL-1α and IL-1β are both approximately 17 kDa with some heterogeneity in the amount of glycosylation.

Physical form

Lyophilized from a 0.2 μm filtered buffered solution.

Analysis Note

The biological activity is measured by the dose-dependent stimulation of MTS cleavage by murine D10S cells.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janette Bester et al.
Scientific reports, 6, 32188-32188 (2016-08-27)
Complex interactions exist between cytokines, and the interleukin family plays a fundamental role in inflammation. Particularly circulating IL-1β, IL-6 and IL-8 are unregulated in systemic and chronic inflammatory conditions. Hypercoagulability is an important hallmark of inflammation, and these cytokines are
Nicholas F Lahens et al.
BMC genomics, 18(1), 602-602 (2017-08-12)
Though Illumina has largely dominated the RNA-Seq field, the simultaneous availability of Ion Torrent has left scientists wondering which platform is most effective for differential gene expression (DGE) analysis. Previous investigations of this question have typically used reference samples derived
Tanja Mang et al.
International journal of molecular sciences, 21(2) (2020-01-18)
The environment surrounding chondrocytes changes drastically in osteoarthritis (OA). For instance, the osmolarity in cartilage (ranging from 350 to 460 mOsm in healthy tissue) decreases during the progression of OA, reaching 270 mOsm. The objective of this study was to
Seong Soo Joo et al.
Biological & pharmaceutical bulletin, 33(11), 1854-1860 (2010-11-05)
In pancreatic islets, free radical formation produced upon exposure to proinflammatory cytokines mediates β cell destruction, which ultimately leads to type 1 diabetes (T1D). In this study, we examined whether laccase, a family of the blue copper protein, can be
Karin Larsson et al.
British journal of pharmacology, 176(24), 4625-4638 (2019-08-14)
Microsomal PGE synthase-1 (mPGES-1), the inducible synthase that catalyses the terminal step in PGE2 biosynthesis, is of high interest as therapeutic target to treat inflammation. Inhibition of mPGES-1 is suggested to be safer than traditional NSAIDs, and recent data demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service