Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9387

Sigma-Aldrich

Indole-3-acetyl-L-aspartic acid

suitable for plant cell culture

Synonym(s):

IAA-L-Asp

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

technique(s)

cell culture | plant: suitable

storage temp.

−20°C

SMILES string

OC(=O)CC(NC(=O)Cc1c[nH]c2ccccc12)C(O)=O

Biochem/physiol Actions

Indole-3-acetyl-L-aspartic acid is a naturally occurring auxin conjugate that regulates free IAA levels in various plant species

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service