Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9386

Sigma-Aldrich

myo-Inositol 1,3,4,6-tetrakis-phosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16O18P4 · xNH3
CAS Number:
Molecular Weight:
500.08 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

N.O[C@H]1[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H16O18P4.H3N/c7-1-3(21-25(9,10)11)5(23-27(15,16)17)2(8)6(24-28(18,19)20)4(1)22-26(12,13)14;/h1-8H,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20);1H3/t1-,2+,3-,4+,5-,6+;

InChI key

ANQVRWQIKKBLLK-PWRMZBPBSA-N

Biochem/physiol Actions

Accumulates with angiotensin II stimulation; stimulates intracellular Ca++ release.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Gawler et al.
The Biochemical journal, 276 ( Pt 1), 163-167 (1991-05-15)
The potential Ca2(+)-releasing activity of the inositol tetrakisphosphates Ins(1,3,4,6)P4 and DL-Ins(1,4,5,6)P4 and the inositol pentakisphosphate Ins(1,3,4,5,6)P5 and their effect on Ins(1,4,5)P3- and DL-Ins (1,3,4,5)P4-mediated Ca2+ release were examined in permeabilized SH-SY5Y human neuroblastoma cells. Neither DL-Ins(1,4,5,6)P4 nor Ins(1,3,4,5,6)P5 exhibit Ca2(+)-releasing
T Balla et al.
Biochemical and biophysical research communications, 148(1), 199-205 (1987-10-14)
Angiotensin II stimulates the formation of several inositol polyphosphates in cultured bovine adrenal glomerulosa cells prelabelled with [3H] inositol. Analysis by high performance anion exchange chromatography of the inositol-phosphate compounds revealed the existence of two additional inositol tetrakisphosphate (InsP4) isomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service