Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I8766

Sigma-Aldrich

5-Iodo-2,4-dimethoxy­pyrimi­dine

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7IN2O2
CAS Number:
Molecular Weight:
266.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

biological source

synthetic (organic)

Assay

≥98% (TLC)

form

solid

mp

71-75 °C (lit.)

storage temp.

−20°C

SMILES string

COc1ncc(I)c(OC)n1

InChI

1S/C6H7IN2O2/c1-10-5-4(7)3-8-6(9-5)11-2/h3H,1-2H3

InChI key

KNTMOGOYRYYUIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kundu, N.G., et al.
Journal of the Chemical Society. Perkin Transactions 1, 1077-1077 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service