Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7409

Sigma-Aldrich

Monoclonal Anti-Interleukin-1α propeptide antibody produced in mouse

clone 409405, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL1, Anti-IL1A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

409405, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL1A(3552)

General description

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced by the activation of NF-κ B transcription factor in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signalling through IL-1 receptor type 1. The IL-1α precursor produced initially is cleaved by calpain to produce a 16 kDa propeptide and 17 kDa mature protein. A variety of cells such as macrophages, epithelial cells, fibroblasts, keratinocytes, monocytes and dendritic cells secrete IL-1α. Both IL-1α and IL-1β are important in acquired immunity, inflammation, response to infection and production of matrix metalloproteinases (MMPs). The propeptide has nuclear localization sequence that enables it to shuttle to nucleus and activate transcription of target genes. The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation.Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis
Monoclonal Anti-Interleukin-1a propeptide recognizes human Interleukin-1α propeptide. It does not cross-react with mature forms of mouse IL-1α or rabbit IL-1α.

Immunogen

purified, E. coli-derived recombinant human interleukin-1α propeptide (aa 1-112).

Application

Anti-interleukin 1α antibody is suitable for immunoblotting at a working concentration of 1-2 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Novak et al.
Scientific reports, 10(1), 6995-6995 (2020-04-26)
Interleukin-1α (IL-1α) is a dual-function proinflammatory mediator. In addition to its role in the canonical IL-1 signaling pathway, which employs membrane-bound receptors, a growing body of evidence shows that IL-1α has some additional intracellular functions. We identified the interaction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service