Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I6146

Sigma-Aldrich

Monoclonal Anti-Interleukin-4 Soluble Receptor antibody produced in mouse

clone 25463.11, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL-4sR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

25463.11, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL4R(3566)

Specificity

The antibody will neutralize human cell surface IL-4 receptor mediated bioactivity.

Immunogen

recombinant human IL soluble receptor produced by the insect cell line Sf 21.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Preparation Note

Purified using protein A

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service