Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5897

Supelco

Magnesium ionophore II

Synonym(s):

ETH 5214, N,N′′-Octamethylene-bis(N′-heptyl-N′-methyl-methylmalonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H62N4O4
CAS Number:
Molecular Weight:
566.86
Beilstein:
6018631
MDL number:
UNSPSC Code:
26111700

storage temp.

2-8°C

SMILES string

CCCCCCCN(C)C(=O)C(C)C(=O)NCCCCCCCCNC(=O)C(C)C(=O)N(C)CCCCCCC

InChI

1S/C32H62N4O4/c1-7-9-11-17-21-25-35(5)31(39)27(3)29(37)33-23-19-15-13-14-16-20-24-34-30(38)28(4)32(40)36(6)26-22-18-12-10-8-2/h27-28H,7-26H2,1-6H3,(H,33,37)(H,34,38)

InChI key

UTFJQHTUZRJEKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service