Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5657

Sigma-Aldrich

Anti-Interleukin-22 Receptor, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IL-22R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~62 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL22RA1(58985)

Immunogen

synthetic peptide corresponding to amino acids 31-46 of human IL-22 recptor precursor.

Biochem/physiol Actions

Interleukin-22 (IL-22) belongs to the IL-10 family of T-cell associated cytokines, highly expressed by Th17 cell subset and also by cells of innate immunity system. The cellular effects of IL-22 are mediated by a heterodimeric receptor made up of IL-22 and IL-10Rβ. IL-22 signaling mediates proliferation of epithelial cells during inflammation mainly by the activation of STAT1 and STAT3 signaling.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren A Zenewicz et al.
European journal of immunology, 38(12), 3265-3268 (2008-11-20)
IL-22 is a Th17 T-cell-associated cytokine that is highly expressed during chronic inflammation. IL-22 receptor expression is absent on immune cells, but is instead restricted to the tissues, providing signaling directionality from the immune system to the tissues. Through Stat3
Shomyseh Sanjabi et al.
Current opinion in pharmacology, 9(4), 447-453 (2009-06-02)
Cytokines play a major role in maintaining lymphocyte homeostasis under both steady-state and inflammatory conditions. Unregulated lymphocytes in steady-state conditions can lead to autoimmunity, whereas during inflammation they can cause excessive tissue damage. Regulatory cytokines function in combination with other
M H Xie et al.
The Journal of biological chemistry, 275(40), 31335-31339 (2000-07-06)
We report the identification of a novel human cytokine, distantly related to interleukin (IL)-10, which we term IL-22. IL-22 is produced by activated T cells. IL-22 is a ligand for CRF2-4, a member of the class II cytokine receptor family.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service