Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5629

Sigma-Aldrich

Iminodiacetic acid

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2COOH)2
CAS Number:
Molecular Weight:
133.10
Beilstein:
878499
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

mp

243 °C (dec.) (lit.)

SMILES string

OC(=O)CNCC(O)=O

InChI

1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)

InChI key

NBZBKCUXIYYUSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service