Skip to Content
MilliporeSigma
All Photos(1)

Documents

I5150

Supelco

ICI 204,448 hydrochloride

Synonym(s):

(±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride, R,S-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl] phenoxy]-acetic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26Cl2N2O4 · HCl
CAS Number:
Molecular Weight:
501.83
MDL number:
UNSPSC Code:
41116107

SMILES string

Cl[H].CN(C(CN1CCCC1)c2cccc(OCC(O)=O)c2)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C23H26Cl2N2O4.ClH/c1-26(22(28)12-16-7-8-19(24)20(25)11-16)21(14-27-9-2-3-10-27)17-5-4-6-18(13-17)31-15-23(29)30;/h4-8,11,13,21H,2-3,9-10,12,14-15H2,1H3,(H,29,30);1H

InChI key

FNDGLVOYAQNQPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Highly selective β2 adrenoceptor antagonist

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Bilski et al.
Journal of cardiovascular pharmacology, 5(3), 430-437 (1983-05-01)
While specific antagonists of the beta 1-adrenoceptor, such as atenolol and betaxolol, are widely available, a potent specific antagonist selective for the beta 2-adrenoceptor has yet to be described. Previously described beta 2-selective antagonists such as butoxamine, H 35/25, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service