Skip to Content
MilliporeSigma
All Photos(1)

Documents

I4881

Sigma-Aldrich

IgG from chicken serum

reagent grade, ≥95% (SDS-PAGE), lyophilized powder

Synonym(s):

Chicken IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

conjugate

unconjugated

grade

reagent grade

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

storage temp.

2-8°C

Physical form

Lyophilized powder containing sodium chloride and sodium phosphate, pH 7.2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Norup et al.
Poultry science, 86(11), 2322-2326 (2007-10-24)
To investigate the ability of chicken mannan-binding lectin (cMBL) to work as a complement activator, a heterogeneous ELISA test was developed, in which the deposition of human complement factor 4 (C4) was used as a measure of complement activation ability.
T G G Groothuis et al.
Journal of evolutionary biology, 19(4), 1304-1313 (2006-06-20)
We investigated in the black-headed gull whether female deposition of antioxidants and immunoglobulins (enhancing early immune function), and testosterone (suppressing immune function and increasing early competitive skills) correlate suggesting that evolution has favoured the mutual adjustment of different pathways for
Marina Kriajevska et al.
The Journal of biological chemistry, 277(7), 5229-5235 (2002-02-12)
Metastasis-associated protein S100A4 (Mts1) induces invasiveness of primary tumors and promotes metastasis. S100A4 belongs to the family of small calcium-binding S100 proteins that are involved in different cellular processes as transducers of calcium signal. S100A4 modulates properties of tumor cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service