Key Documents
I4778
Sigma-Aldrich
Igloo® cooler ice bucket
About This Item
UNSPSC Code:
41103000
manufacturer/tradename
Jencons
capacity
3.5 L
external H × W × D
280 mm × 300 mm × 200 mm
General description
Specially designed to provide a rugged container for long-term thermal storage of ice and dry ice on the bench. Smooth, easy-to-clean, chemically resistant white polystyrene body and lid have a 25 mm thick double insulated wall filled with expanded polystyrene foam. Made with four convenient support holes to accept your single channel pipettors.
Legal Information
Igloo is a registered trademark of Igloo Products Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)