Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4026

Sigma-Aldrich

Interleukin-17 from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

CTLA-8, IL-17

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

16.5 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il17a(16171)

Biochem/physiol Actions

IL-17 (interleukin-17) participates in the activation of nuclear factor-κB and mitogen-activated protein kinase pathways and CCAAT/enhancer-binding protein family of transcription factors, thereby regulating pro-inflammatory and antimicrobial genes. In rheumatoid arthritis, IL-17 is responsible for the inflammation as well as destruction of joints.
Initially known as CTLA-8; is a T cell-expressed polypeptide that appears to be involved in the regulation of the hematopoietic system; has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis.
Interleukin 17 (IL-17), initially known as CTLA-8, is a T cell-expressed polypeptide that appears to be involved in the regulation of the hematopoietic system. IL-17 has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 1.25 mg of bovine serum albumin.

Analysis Note

The biological activity of mouse IL-17 is measured by its ability to induce IL-6 production using NIH/3T3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoki Maekawa et al.
Nature communications, 6, 8272-8272 (2015-09-17)
Del-1 is an endothelial cell-secreted anti-inflammatory protein. In humans and mice, Del-1 expression is inversely related to that of IL-17, which inhibits Del-1 through hitherto unidentified mechanism(s). Here we show that IL-17 downregulates human endothelial cell expression of Del-1 by
Annica Andersson et al.
Clinical immunology (Orlando, Fla.), 161(2), 324-332 (2015-10-02)
Interleukin-17 (IL-17) drives inflammation and destruction of joints in rheumatoid arthritis (RA). The female sex hormone 17β-estradiol (E2) inhibits experimental arthritis. γδT cells are significant producers of IL-17, thus the aim of this study was to investigate if E2 influenced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service