Skip to Content
MilliporeSigma
All Photos(1)

Documents

I2259

Sigma-Aldrich

myo-Inositol hexanicotinate

Synonym(s):

myo-Inositol hexa-3-pyridinecarboxylate, Inositol niacinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H30N6O12
CAS Number:
Molecular Weight:
810.72
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

mp

254-256 °C (lit.)

SMILES string

O=C(O[C@H]1[C@@H](OC(=O)c2cccnc2)[C@H](OC(=O)c3cccnc3)[C@@H](OC(=O)c4cccnc4)[C@H](OC(=O)c5cccnc5)[C@H]1OC(=O)c6cccnc6)c7cccnc7

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Does inositol nicotinate (Hexopal) influence intermittent claudication? A controlled trial.
R S Kiff et al.
The British journal of clinical practice, 42(4), 141-145 (1988-04-01)
The therapeutic efficacy of inositol nicotinate (Hexopal) in intermittent claudication: a controlled trial.
J O'Hara et al.
The British journal of clinical practice, 42(9), 377-383 (1988-09-01)
Treatment of intermittent claudication with inositol nicotinate.
A Head
The Practitioner, 230(1411), 49-54 (1986-01-01)
L Harthon et al.
Arzneimittel-Forschung, 29(12), 1859-1862 (1979-01-01)
The esters between nicotinic acid and pentaerythritol (niceritrol, Perycit) or meso-inositol (MIHN) were hydrolyzed by enzymes present in plasma and tissues of man, rat and dog. Human plasma and rat tissues hydrolyzed niceritrol at a higher rate than did MIHN.
Yu Pengzhan et al.
Pharmacological research, 48(6), 543-549 (2003-10-07)
Ulvan, a sulfated polysaccharide from Ulva pertusa, was degraded to yield two low molecular weight fractions U1 and U2. The molecular weights of ulvan and its fractions were determined and varied from 151.6 to 28.2 kDa. They were fed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service