Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2257

Sigma-Aldrich

N-(2-Iodoethyl)trifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5F3INO
CAS Number:
Molecular Weight:
266.99
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)NCCI

Application

Reagent for aminoethylation of sulfhydryl groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-(beta-Iodoethyl)trifluoroacetamide: a new reagent for the aminoethylation of thio groups in proteins.
W E Schwartz et al.
Analytical biochemistry, 106(1), 43-48 (1980-07-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service