Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1909

Sigma-Aldrich

Immethridine hydrobromide

≥98% (HPLC), solid

Synonym(s):

4-(1H-Imidazol-4-ylmethyl)pyridine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3 · xHBr
CAS Number:
Molecular Weight:
159.19 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: ~10 mg/mL
H2O: insoluble

SMILES string

Br[H].C(c1ccncc1)c2c[nH]cn2

InChI

1S/C9H9N3.BrH/c1-3-10-4-2-8(1)5-9-6-11-7-12-9;/h1-4,6-7H,5H2,(H,11,12);1H

InChI key

XAOXYOKDVRPLBH-UHFFFAOYSA-N

Biochem/physiol Actions

Potent, highly selective H3 histamine receptor agonist

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruengwit Kitbunnadaj et al.
Journal of medicinal chemistry, 47(10), 2414-2417 (2004-04-30)
In this study, the piperidine ring of immepip and its analogues was replaced by a rigid heterocyclic pyridine ring. Many compounds in the series exhibit high affinity and agonist activity at the human histamine H(3) receptor. Particularly, the 4-pyridinyl analogue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service