Skip to Content
MilliporeSigma
All Photos(1)

Documents

I148

Sigma-Aldrich

Imperatoxin A

>88%, lyophilized powder

Synonym(s):

Ip Txa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C148H260N58O45S6
CAS Number:
Molecular Weight:
3764.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

>88%

form

lyophilized powder

storage temp.

−20°C

Amino Acid Sequence

Gly-Asp-Cys-Leu-Pro-His-Leu-Lys-Arg-Cys-Lys-Ala-Asp-Asn-Asp-Cys-Cys-Gly-Lys-Lys-Cys-Lys-Arg-Arg-Gly-Thr-Asn-Ala-Glu-Lys-Arg-Cys-Arg

Biochem/physiol Actions

Selective skeletal-type ryanodine sensitive intracellular Ca2+ channel activator found in the scorpion Pandinus

Features and Benefits

This compound is featured on the InsP3/Ryanodine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Z Zamudio et al.
FEBS letters, 405(3), 385-389 (1997-04-01)
We present the complete amino acid sequence of Imperatoxin A (IpTx(a)), a 33-amino-acid peptide from the venom of the scorpion P. imperator which activates Ca2+ release channels/ryanodine receptors (RyR) of sarcoplasmic reticulum (SR). The amino acid sequence of IpTx(a) shows
H H Valdivia et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(24), 12185-12189 (1992-12-15)
We report the purification of two peptides, called "imperatoxin inhibitor" and "imperatoxin activator," from the venom of the scorpion Pandinus imperator targeted against ryanodine receptor Ca(2+)-release channels. Imperatoxin inhibitor has a M(r) of approximately 10,500, inhibits [3H]ryanodine binding to skeletal
R el-Hayek et al.
The Journal of biological chemistry, 270(48), 28696-28704 (1995-12-01)
We have used [3H]ryanodine binding experiments and single channel recordings to provide convergent descriptions of the effect of imperatoxin A (IpTxa), a approximately 5-kDa peptide from the venom of the scorpion Pandinus imperator (Valdivia, H. H., Kirby, M. S., Lederer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service