Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1007

Sigma-Aldrich

6-(Iodoacetamido)hexanoic acid N-hydroxysuccinimide ester

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17IN2O5
CAS Number:
Molecular Weight:
396.18
MDL number:
UNSPSC Code:
12171500

form

powder

solubility

methanol: 50 mg/mL
DMF: soluble

storage temp.

−20°C

SMILES string

ICC(=O)NCCCCCC(=O)ON1C(=O)CCC1=O

Application

A heterobifunctional cross-linking reagent containing an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5) Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 7.5 (7.0-8.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates. Provides a 9-atom linker.

Caution

The iodoacetyl group is light sensitive.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E S Rector et al.
Journal of immunological methods, 24(3-4), 321-336 (1978-01-01)
A novel procedure for the synthesis of well-defined protein-protein conjugates is described using ovalbumin (OA) and IgG as test proteins. This procedure involves the highly selective and rapid reaction of alkyl halide and sulfhydryl groups, which have been grafted, respectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service