Key Documents
HS8881D
Vinyl-Coated Lead Ring ("C" shape)
fits beaker size, 1000 to 4000 mL, blue
Select a Size
1 EACH
$55.90
$55.90
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$55.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.71
$55.90
Please contact Customer Service for Availability
material
blue
feature
fits beaker size 1000 to 4000 mL
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS8881D
General description
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.
Features and Benefits
  • Vinyl-coated lead ring will not mark or scratch benchtops and labware
  • O-Shape Style
Choose from one of the most recent versions:
Certificates of Analysis (COA)