Documents
HS120686
Clinical 4-Way Tube Rack
green
About This Item
UNSPSC Code:
41122800
NACRES:
NB.17
material
green
polypropylene
feature
to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific 120686
L × W × H
232 mm × 70 mm × 95 mm
General description
The Clinical 4-Way Tube Rack is the only tube rack specifically designed to hold round, flat & conical blood collection and culture tubes. It holds multiple tube sizes without multiple racks.
Features and Benefits
  • Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection & culture tubes
  • Only 4-way rack available with a six way connection, link different or the same well sizes, end to end or side by side, in any orientation
  • Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Robust, made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezers
  • Assorted color pack available for color coding options
  • Patent Pending
  • Autoclavable