Documents
HS120644
Reagent Reservoir
capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (packed as 5 ea)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
material
V-bottom bottom
polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 100 (packed as 5 ea)
manufacturer/tradename
Heathrow Scientific 120644
capacity
50 mL
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending