Documents
HS120627
smartPans Cahn Style Aluminum Microbalance Weigh Pan
pan diam. 25 mm
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
material
aluminum
packaging
pack of 20 ea
manufacturer/tradename
Heathrow Scientific 120627
H
3.6 mm
pan diam.
25 mm
General description
The Cahn style aluminum weigh pan is ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
Features and Benefits
  • Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
  • Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil
  • Foil meets ASTM B-479, specification 3.1.4 and 10.3.1
  • Cahn style” accessory pans are compatible with most commercially available analyzers and microbalances
  • Pan is recyclable
Legal Information
smartPans is a trademark of LevGo Inc.