Key Documents
HS120222
Weigh Boat
Small, square blue polystyrene, Anti-static, pk of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Select a Size
500 EA
$62.10
$62.10
Please contact Customer Service for Availability
Select a Size
Change View
500 EA
$62.10
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
$62.10
Please contact Customer Service for Availability
material
square blue polystyrene
size
small
feature
Anti-static
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS120222
L × W × H
(46 mm) × 46 mm × 8 mm
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer
Choose from one of the most recent versions:
Certificates of Analysis (COA)