Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA059739

Sigma-Aldrich

Anti-FAP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DPPIV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

ERCQYYTASFSDYAKYYALVCYGPGIPISTLHDGRTDQEIKILEENKELENALKNIQLPKEEIKKLEVDEITLWYKM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FAP(2191)

General description

Fibroblast activation protein (FAP) is encoded by the gene mapped to human chromosome 2q24.2. It is a type II transmembrane serine protease and a cell surface antigen. FAP is a homodimeric integral protein with dipeptidyl peptidase IV like fold. The protein is characterized with an α/β-hydrolase domain and an eight-bladed β-propeller domain. It is not expressed in normal tissues. FAP is only expressed by activated fibroblasts in response to pathologic situations.

Immunogen

fibroblast activation protein, alpha

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collecation of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Anti-FAP antibody produced in rabbit has been used in immunohistochemical staining.

Biochem/physiol Actions

Fibroblast activation protein (FAP) exhibits in vitro dipeptidyl peptidase activity and collagenolytic activity. It cleaves N-terminal dipeptides from polypeptides and can degrade gelatin and type I collagen. FAP is expressed in several pathogenic sites including cancer, fibrosis, arthritis, wounding, or inflammation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86260.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibroblasts in the Tumor Microenvironment: Shield or Spear?.
Alkasalias T, et al.
International Journal of Molecular Sciences, 19(5), 1532-1532 (2018)
Striking similarities in genetic aberrations between a rectal tumor and its lung recurrence.
Rahma O E, et al.
World Journal of Gastrointestinal Oncology, 5(11), 198?203-198?203 (2013)
Fibroblast activation protein, a dual specificity serine protease expressed in reactive human tumor stromal fibroblasts.
Park J E, et al.
The Journal of Biological Chemistry, 274(51), 36505-36512 (1999)
Thomas Kelly et al.
International review of cell and molecular biology, 297, 83-116 (2012-05-23)
Fibroblast activation protein-α (FAP) is a serine protease that can provide target specificity to therapeutic agents because in adults its expression is restricted to pathologic sites, including cancer, fibrosis, arthritis, wounding, or inflammation. It is not expressed in most normal
Kathleen Aertgeerts et al.
The Journal of biological chemistry, 280(20), 19441-19444 (2005-04-06)
Fibroblast activation protein alpha (FAPalpha) is highly expressed in epithelial cancers and has been implicated in extracellular matrix remodeling, tumor growth, and metastasis. We present the first high resolution structure for the apoenzyme as well as kinetic data toward small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service