Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA048938

Sigma-Aldrich

Anti-SLC9A7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NHE7, Anti-solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

QVYDNQEPLREEDSDFILTEGDLTLTYGDSTVTANGSSSSHTASTSLEGSRRT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC9A7(84679)

General description

Solute carrier family 9 member 7 (SLC9A7), also called sodium/hydrogen exchanger 7 (NHE7), is a 80 kDa protein present in trans-Golgi complex. It is a sodium/hydrogen exchanger with N-terminal membrane spanning region and C-terminal hydrophilic tail. SLC9A7 is mapped to human chromosome Xp11.3.

Immunogen

solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 9 member 7 (SLC9A7) maintains cation homeostasis in trans-Golgi. Its interaction with secretory carrier membrane protein (SCAMP) is crucial for regulating transport in the trans-Golgi complex. SLC9A7 functions like vacuolar (H+)-ATPases (V-ATPases), as a proton-loading transporter. SLC9A7 is highly is expressed in breast cancer and contributes to tumor progression. Polymorphisms in the SLC9A7 gene may contribute to pathogenesis of Alzheimer′s disease. Missense mutation in SLC9A7 is also implicated with metabolic disorder,cystinuria. Deletion of SLC9A7 gene is implicated X-linked disorder, resulting in mental retardation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84427

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZNF674: a new kruppel-associated box-containing zinc-finger gene involved in nonsyndromic X-linked mental retardation
Lugtenberg D, et al.
American Journal of Human Genetics, 78(2), 265-278 (2006)
The intracellular Na+/H+ exchanger NHE7 effects a Na+-coupled, but not K+-coupled proton-loading mechanism in endocytosis
Milosavljevic N, et al.
Cell Reports, 7(3), 689-696 (2014)
Delineation of cystinuria in Saudi Arabia: A case series
Obaid A, et al.
BMC Nephrology, 18(1), 50-50 (2017)
Secretory carrier membrane proteins interact and regulate trafficking of the organellar (Na+, K+)/H+ exchanger NHE7
Lin PJC, et al.
Journal of Cell Science, 118(9), 1885-1897 (2005)
Organellar (Na+, K+)/H+ exchanger NHE7 regulates cell adhesion, invasion and anchorage-independent growth of breast cancer MDA-MB-231 cells
Onishi I, et al.
Oncology Reports, 27(2), 311-317 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service