Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA047509

Sigma-Aldrich

Anti-TMEM176B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LR8, Anti-transmembrane protein 176B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VGLRNLCGQSSQPLNEEGSEKRLLGENSVPPSPSREQTSTAIVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

TMEM176B (transmembrane protein 176B) is a member of MS4A (membrane-spanning 4-domain family, subfamily A) family of proteins. Its molecular weight is 29.05kDa and is also known as LR8 (transmembrane protein). Its transcripts are more in liver, kidney and lung. TMEM176B codes for a non selective monovalent cation channel, that is triggered by acidification. It is located on chromosome 7q36.1.

Immunogen

transmembrane protein 176B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TMEM176B (transmembrane protein 176B) plays a major role in the activation and tolerance of immune cell. It also plays an important role in the maturation process of dendritic cells (DCs).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85038.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The MS4A family: counting past 1, 2 and 3.
Eon Kuek L, et al.
Immunology and Cell Biology, 94(1), 11-23 (2016)
ROR?t+ cells selectively express redundant cation channels linked to the Golgi apparatus.
Drujont L, et al.
Scientific Reports, 6:23682 (2016)
Tmem176B and Tmem176A are associated with the immature state of dendritic cells.
Condamine T, et al.
Journal of Leukocyte Biology, 88(3), 507-515 (2010)
Math P Cuajungco et al.
Acta histochemica, 114(7), 705-712 (2012-01-17)
Transmembrane (TMEM)-176A and 176B proteins belong to the MS4A family of proteins whose function in the immune system remains unclear. TMEM176A transcripts were previously shown to be elevated in liver cancer or kidney tissue with proteinuria, while marked changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service