Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA043237

Sigma-Aldrich

Anti-APOBEC3A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apolipoprotein b mrna editing enzyme, catalytic polypeptide-like 3a, Anti-Arp3, Anti-Phrbn

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000-1:2500
western blot: 0.04-0.4 μg/mL

immunogen sequence

EASPASGPRHLMDPHIFTSNFNNGIGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene APOBEC3A (apolipoprotein B mRNA editing enzyme catalytic subunit 3A) is mapped to human chromosome 22q13.1. The protein has a catalytically active deaminase domain. It belongs to the APOBEC3 genes family.

Immunogen

apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-APOBEC3A antibody produced in rabbit has been used in immunohistochemistry.
Anti-APOBEC3A antibody produced in rabbit has also been used in immunofluorescence staining.

Biochem/physiol Actions

APOBEC3A (apolipoprotein B mRNA editing enzyme catalytic subunit 3A) is a cytidine deaminase. APOBEC3 proteins are mainly involved in intracellular antiviral defense responses, particularly against retroviruses. They are also involved in carcinogenesis by dC (deoxycytidine) deamination, thereby causing DNA mutations. APOBEC3A also exhibits mRNA-editing function. It is involved with guanine-to-alanine editing in transcripts of WT1 (wilms tumor 1). Deletion in APOBEC3A gene is associated with breast cancer predisposition. This happens due to increased stability of APOBEC3A mRNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83236

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of functional germline variants in APOBEC3 and driver genes on breast cancer risk in Moroccan study population.
Marouf C, et al.
BMC Cancer, 16, 165-165 (2016)
APOBEC3A is implicated in a novel class of G-to-A mRNA editing in WT1 transcripts.
Niavarani A, et al.
PLoS ONE, 10, e0120089-e0120089 (2015)
Makoto Kano et al.
International journal of cancer, 145(6), 1547-1557 (2019-06-23)
Human papillomavirus (HPV) has been identified as a causative agent of cervical cancer and oropharyngeal cancer (OPC). Intriguingly, estrogen and HPV were shown to play synergistic roles in cervical carcinogenesis. We recently demonstrated that the apolipoprotein B mRNA-editing catalytic polypeptide
The AID/APOBEC family of nucleic acid mutators.
Conticello SG
Genome Biology, 229, doi: 10- doi: 10 (2008)
APOBEC3A associates with human papillomavirus genome integration in oropharyngeal cancers.
Kondo S, et al.
Oncogene, doi: 10-doi: 10 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service