Skip to Content
MilliporeSigma
All Photos(8)

Documents

HPA027452

Sigma-Aldrich

Anti-PPP1R8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3

Synonym(s):

Anti-ARD-1, Anti-NIPP-1, Anti-Nuclear inhibitor of protein phosphatase 1, Anti-Protein phosphatase 1 regulatory inhibitor subunit 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TFLGHIRLEPHKPQQIPIDSTVSFGASTRAYTLREKPQTLPSAVKGDEKMGGEDDELKGLLGLPEEETELDNLTEFNTAHNK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1R8(5511)

General description

The gene PPP1R8 (protein phosphatase 1 regulatory inhibitor subunit 8) is mapped to human chromosome 1p35. The encoded protein has a PP1 (protein phosphatase 1)-anchoring domain, a forkhead-associated (FHA) domain and a PP1-inhibitory domain. PPP1R8 is widely expressed.

Immunogen

Nuclear inhibitor of protein phosphatase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PPP1R8 (protein phosphatase 1 regulatory inhibitor subunit 8) forms a heterodimer with nuclear PP1 (protein phosphatase 1) and works as an inhibitor of PP1. It is also involved in spliceosome arrangement and transcriptional silencing by the histone methyltransferase EZH2 (enhancer of zeste homolog 2). The protein also enhances Cdc42 (cell division control protein 42 homolog)-associated migration of HeLa cancer cells. Absence of PPP1R8 gene in mice results in embryonic lethality.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76662

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nele Van Dessel et al.
FEBS letters, 589(12), 1314-1321 (2015-04-25)
The deletion of the protein phosphatase-1 (PP1) regulator known as Nuclear Inhibitor of PP1 (NIPP1) is embryonic lethal during gastrulation, hinting at a key role of PP1-NIPP1 in lineage specification. Consistent with this notion we show here that a mild
Organization and alternate splice products of the gene encoding nuclear inhibitor of protein phosphatase-1 (NIPP-1).
Van Eynde A, et al.
European Journal of Biochemistry, 261, 291-300 (1999)
The molecular basis for substrate specificity of the nuclear NIPP1:PP1 holoenzyme.
O'Connell N, et al.
Structure, 20, 1746-1756 (2012)
Shunsuke Hanaki et al.
Cancer science, 112(7), 2739-2752 (2021-05-04)
DNA damage induces transcriptional repression of E2F1 target genes and a reduction in histone H3-Thr11 phosphorylation (H3-pThr11 ) at E2F1 target gene promoters. Dephosphorylation of H3-pThr11 is partly mediated by Chk1 kinase and protein phosphatase 1γ (PP1γ) phosphatase. Here, we
Mónica Ferreira et al.
The Journal of biological chemistry, 293(47), 18031-18039 (2018-10-12)
Germ cell proliferation is epigenetically controlled, mainly through DNA methylation and histone modifications. However, the pivotal epigenetic regulators of germ cell self-renewal and differentiation in postnatal testis are still poorly defined. The histone methyltransferase enhancer of zeste homolog 2 (EZH2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service