Skip to Content
MilliporeSigma
All Photos(6)

Documents

HPA026871

Sigma-Aldrich

Anti-STAB2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-190 kDa hyaluronan receptor for endocytosis, Anti-FAS1 EGF-like and X-link domain-containing adhesion molecule 2, Anti-FEEL-2, Anti-Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 2, Anti-Hyaluronan receptor for endocytosis, Anti-Stabilin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VVLEEKLLKNDLHNGMHRETMLGFSYFLSFFLHNDQLYVNEAPINYTNVATDKGVIHGLGKVLEIQKNRCDNNDTTIIRGRCRTCSSELTCPFGTKSLGNEKRRCIYTSYFMGRRTLFIGCQPKCVRTVITR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAB2(55576)

General description

The Stabilin-2 (STAB2) gene with 69 exons spanning 180kbp is mapped to human chromosome 12. Stabilin-2, also known as fasciclin, EGF-like, laminin-type EGF-like, and link domain-containing scavenger receptor-2 (FEEL-2) or hyaluronan receptor for endocytosis (HARE), is a large receptor containing 2551 amino acids belonging to the family of fasciclin-like hyaluronan receptor homologues. STAB2 transcript is expressed in spleen, bone marrow and liver.

Immunogen

Stabilin-2 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The hyaluronan receptor for endocytosis (HARE) or stabilin-2 functions as a mammalian endocytic clearance receptor for hyaluronic acid (HA), heparin, advanced glycation end-products, acetylated and oxidized low-density lipoproteins and collagen N-terminal propeptides. HARE/Stab2 facilitates binding of lymphocyte to sinusoidal endothelium by interacting with αMβ2 and α5β5 integrins. The encoded protein mediates phagocytosis of bacteria and apoptotic cells displaying cell surface phosphatidylserine. In mice, Stab-2 might assist in separating hyaluronan from the lymph or the blood circulation through the network of endothelial sinuses. The protein also exhibits scavenger receptor activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71958

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FEEL-1, a novel scavenger receptor with in vitro bacteria-binding and angiogenesis-modulating activities.
Adachi H, Tsujimoto M
The Journal of Biological Chemistry, 277(37), 34264-34270 (2002)
Expression of stabilin-2, a novel fasciclin-like hyaluronan receptor protein, in murine sinusoidal endothelia, avascular tissues, and at solid/liquid interfaces.
Falkowski M
Histochemistry and Cell Biology, 120(5), 361-369 (2003)
Tingting Bian et al.
Oncotarget, 8(38), 63442-63452 (2017-10-04)
Pulmonary enteric adenocarcinoma (PEAC), a rare type of non-small cell lung cancer, has similar histological and immunohistochemical morphology to colorectal adenocarcinoma. Cadherin-17 (CDH17) and SATB homeobox 2 (SATB2) immunoexpression have recently been demonstrated in colorectal adenocarcinoma. In this study, we
Amanda K Hare et al.
Biochemical and biophysical research communications, 456(1), 257-261 (2014-12-03)
The hyaluronan receptor for endocytosis (HARE), or Stabilin-2, is the mammalian endocytic clearance receptor for HA, heparin, advanced glycation end-products, acetylated and oxidized low-density lipoproteins and collagen N-terminal propeptides. This large 2551 amino acid receptor is encoded by a gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service