Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA023973

Sigma-Aldrich

Anti-EIF3E antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Eukaryotic translation initiation factor 3 subunit 6, Anti-Eukaryotic translation initiation factor 3 subunit E, Anti-Viral integration site protein INT-6 homolog, Anti-eIF-3 p48, Anti-eIF3e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ARLFIFETFCRIHQCISINMLADKLNMTPEEAERWIVNLIRNARLDAKIDSKLGHVVMGNNAVSPYQQVIEKTKSLSFRSQMLAMNIEKKLNQNSRSEAPNWATQDSGFY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF3E(3646)

General description

KRT15 (keratin 15) is a structural protein which is a member of the acidic or type I keratin family. It is a cytoskeletal protein, which pairs with its basic or type II partner, K5 to assemble into a keratin filament network. It is a phenotypic marker for hair follicle bulge, skin keratinocyte, and limbal epithelial stem cells. This protein is expressed by basal layers of stratified epithelium. This gene is localized to human chromosome 17.

Immunogen

Eukaryotic translation initiation factor 3 subunit E recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KRT15 (keratin 15) is a component of the intermediate filament network of the basal layer of stratified epithelia, and mutation in this gene results in epidermolysis bullosa simplex (EBS). This disorder is characterized by basal layer blistering due to cell fragility. The expression of this protein is elevated by interferon (IFN)-γ and thyroid hormone, and thus, they have potential as therapeutic agents for EBS. This protein is also expressed in a population of high-risk breast carcinomas. The expression pattern of this protein might be useful in differentiating trichoepithelioma (TE) from basal cell carcinoma (BCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74683

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chong Won Choi et al.
The Journal of dermatology, 35(8), 499-502 (2008-09-16)
Trichoepithelioma (TE) is a benign neoplasm of the skin that resembles basal cell carcinoma (BCC) in its clinical and histological features. In this study, we evaluate the usefulness of elastic fiber staining and cytokeratin 15 expression pattern in terms of
Nada Radoja et al.
Molecular and cellular biology, 24(8), 3168-3179 (2004-04-03)
Basal layers of stratified epithelia express keratins K5, K14, and K15, which assemble into intermediate filament networks. Mutations in K5 or K14 genes cause epidermolysis bullosa simplex (EBS), a disorder with blistering in the basal layer due to cell fragility.
Julio E Celis et al.
Molecular oncology, 1(3), 321-349 (2007-12-01)
Recently, we presented evidence--based on the analysis of benign hyperproliferative lesions of the breast--for the presence of cells that express the stem cell marker cytokeratin (CK) 15 in combination with CK19, a protein widely expressed by mammary epithelial cells. Here
Philip V'kovski et al.
eLife, 8 (2019-01-12)
Positive-sense RNA viruses hijack intracellular membranes that provide niches for viral RNA synthesis and a platform for interactions with host proteins. However, little is known about host factors at the interface between replicase complexes and the host cytoplasm. We engineered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service