Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA023923

Sigma-Aldrich

Anti-PRR11 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Proline-rich protein 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LAPVLLRKPSLAKALQAGPLKKDGPMQITVKDLLTVKLKKTQSLDEKRKLIPSPKARNPLVTVSDLQHVTLKPNSKVLSTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRR11(55771)

General description

PRR11 (proline rich 11) is a cancer-related gene which was first identified in 2013. The gene is mapped to human chromosome 17q22.

Immunogen

Proline-rich protein 11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRR11 (proline rich 11) is a putative candidate oncogene, which is involved in the tumorigenesis of lung cancer and EMT (epithelial-to-mesenchymal transition) in breast cancer. In breast cancer cells, it reduces the expression of E-cadherin, an epithelial marker, and elevates the expression of mesenchymal markers fibronectin and N-cadherin. It is also up-regulated in gastric cancer (GC), and is thought to promote development and progression of GC. This gene plays an important role in cell cycle progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75526

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zongchang Song et al.
PloS one, 10(8), e0128943-e0128943 (2015-08-08)
PRR11 is a potential candidate oncogene that has been implicated in the pathogenesis of lung cancer, however the role of PRR11 in gastric cancer is currently unclear. In the present study, we investigated the role of PRR11 in gastric cancer
Zhili Wei et al.
Genes & diseases, 9(1), 230-244 (2022-01-11)
Filopodia, a finger-like structure and actin-rich plasma-membrane protrusion at the leading edge of the cell, has important roles in cell motility. However, the mechanisms of filopodia generation are not well-understood via the actin-related protein 2/3 (ARP2/3) complex in Non-Small Cell
Chundong Zhang et al.
Biochemical and biophysical research communications, 458(3), 501-508 (2015-02-11)
Recently, we have demonstrated that proline-rich protein 11 (PRR11) is a novel tumor-related gene product likely implicated in the regulation of cell cycle progression as well as lung cancer development. However, its precise role in cell cycle progression remains unclear.
Yu Zhan et al.
World journal of surgical oncology, 18(1), 318-318 (2020-12-06)
The proline-rich protein 11 (PRR11) is a newly identified oncogene associated with a poor prognosis in several human cancers. Nonetheless, research on its role in ovarian cancer (OC) remains largely understudied. Therefore, this study aims to evaluate the expression levels
Chao Cai et al.
Oncology reports, 39(4), 1658-1670 (2018-02-03)
hsa-miR-195-5p (miR-195) has been proven to be a critical regulator in the progression of prostate cancer (PCa). To identify additional targets and molecular functions of miR-195, we overexpressed miR-195 by transient oligonucleotide transfection in DU145 and LNCaP cells and examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service