Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA023322

Sigma-Aldrich

Anti-MARCH10 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Membrane-associated RING finger protein 10, Anti-Probable E3 ubiquitin-protein ligase MARCH10, Anti-RING finger protein 190

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LPDRESATEKDRGGSENAKKSPLSWDTKSEPRQEVGVNAENVWSDCISVEHRPGTHDSEGYWKDYLNSSQNSLDYFISGRPISPRSSVNSSYNPPAS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

MARCH10 (membrane associated ring-CH-type finger 10) belongs to the MARCH family of proteins. It is a testis-specific gene. The gene is mapped to human chromosome 17q23.

Immunogen

Probable E3 ubiquitin-protein ligase MARCH10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MARCH10 (membrane associated ring-CH-type finger 10) is an E3 ubiquitin ligase which associates with microtubules. It participates in the assembly and maintenance of spermatozoa flagella.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75818

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojun Hou et al.
BMC genomics, 13, 495-495 (2012-09-21)
Ubiquitin-mediated protein modification and degradation are believed to play important roles in mammalian spermatogenesis. The catalogues of ubiquitin activating enzymes, conjugating enzymes, and ligases (E3s) have been known for mammals such as mice and humans. However, a systematic characterization of
Nobuhiro Nakamura
Membranes, 1(4), 354-393 (2011-01-01)
A large number of RING finger (RNF) proteins are present in eukaryotic cells and the majority of them are believed to act as E3 ubiquitin ligases. In humans, 49 RNF proteins are predicted to contain transmembrane domains, several of which
FuJun Liu et al.
Biological & pharmaceutical bulletin, 34(1), 28-35 (2011-01-08)
The testis is the special male gonad responsible for spermatogenesis and steroidogenesis with complex gene expressions. Characterizing and comparing the testis-specific genes in different species can reveal key genes related to testis specific functions and provide supplementary information for study
Tetsuro Yoshida et al.
International journal of molecular medicine, 24(4), 539-547 (2009-09-03)
Chronic kidney disease (CKD) is recognized as a risk factor not only for end-stage renal disease but also for cardiovascular disease. Early detection and treatment of CKD is a likely key factor for prevention of its complications. Although genetic linkage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service