Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA023225

Sigma-Aldrich

Anti-RPS6KA4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-90 kDa ribosomal protein S6 kinase 4, Anti-Nuclear mitogen- and stress-activated protein kinase 2, Anti-RSKB, Anti-Ribosomal protein S6 kinase alpha-4, Anti-Ribosomal protein kinase B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$827.00

$827.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$827.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$827.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

AQEVRNHPFFQGLDWVALAARKIPAPFRPQIRSELDVGNFAEEFTRLEPVYSPPGSPPPGDPRIFQGYSFVAPSILFDHNNAVMTDGLEAPGAGDRPGRAAVARSAMMQDSPFFQQYELDLREPALGQGSFSVC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KA4(8986)

General description

The gene RPS6KA4 (ribosomal protein S6 kinase A4) is mapped to human chromosome 11q. The protein is present in the nucleus. It is a downstream-activated protein of the ERK (extracellular signal-regulated kinase)- and p38-mitogen activated protein kinase (MAPK) signaling pathways.[1]

Immunogen

Ribosomal protein S6 kinase alpha-4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPS6KA4 (ribosomal protein S6 kinase A4) participates in cell survival, proliferation as well as differentiation. It also plays an important role in tumor formation. RPS6KA4 is responsible for the nucleosomal response by phosphorylation of the histone H3 and HMGN1 (high mobility group nucleosome binding domain 1). It is involved in CREB (cAMP responsive element binding protein) activation.[1] It is also responsible for the UV-mediated phosphorylation of p65, which enhances nuclear factor κB activity in MDA-MB-231 (human breast adenocarcinoma) cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73522

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Un Kang et al.
International journal of molecular medicine, 29(1), 81-87 (2011-10-14)
The lymph node metastatic (LNM) spread of tumor cells is a frequent event in the initial process of cancer dissemination and is a powerful independent prognostic indicator in gastric adenocarcinoma (GAC). High density genomic arrays were conducted to identify molecular
Kellie A Jacks et al.
The Journal of biological chemistry, 285(3), 1661-1670 (2009-11-26)
Mitogen- and stress-activated protein kinases, MSK1 and the closely related isoform MSK2, are nuclear kinases that are activated following mitogen stimulation or cellular stress, including UV radiation, by the ERK1/2 and p38 MAPK signaling cascades, respectively. However, factors that differentially
Protiti Khan et al.
PloS one, 8(5), e63189-e63189 (2013-05-16)
Mitogen- and stress-activated protein kinases 1 and 2 (MSK1 and MSK2), activated downstream of the ERK- and p38-mitogen-activated protein kinase pathways are involved in cell survival, proliferation and differentiation. Following mitogenic or stress stimuli, they mediate the nucleosomal response, which
B Pierrat et al.
The Journal of biological chemistry, 273(45), 29661-29671 (1998-10-29)
A novel ribosomal S6 kinase (RSK) family member, RSK-B, was identified in a p38alphaMAPK-baited intracellular interaction screen. RSK-B presents two catalytic domains typical for the RSK family. The protein kinase C-like N-terminal and the calcium/calmodulin kinase-like C-terminal domains both contain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service