Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA022971

Sigma-Aldrich

Anti-GRHPR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Glyoxylate reductase/hydroxypyruvate reductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GDVVNQDDLYQALASGKIAAAGLDVTSPEPLPTNHPLLTLKNCVILPHIGSATHRTRNTMSLLAANNLLAGLRGEPMPSELKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRHPR(9380)

General description

The gene GRHPR (glyoxylate reductase/hydroxypyruvate reductase) is mapped to human chromosome 9. The protein contains 328 amino acids.

Immunogen

Glyoxylate reductase/hydroxypyruvate reductase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GRHPR (glyoxylate reductase/hydroxypyruvate reductase) is a liver enzyme with dual role. It converts glyoxylate to glycolate and hydroxypyruvate to D-glycerate. Mutations in GRHPR or its deficiency are associated with primary hyperoxaluria type 2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75289

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Takayama et al.
Clinical genetics, 86(4), 342-348 (2013-10-15)
The objective of this study was to investigate ethnic differences in the glyoxylate reductase/hydroxypyruvate reductase (GRHPR) gene in patients with primary hyperoxaluria type 2 (PH2). GRHPR was genotyped in Japanese patients with PH2 and all GRHPR mutations described to date
Yinglian Pan et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 80(3), 155-162 (2013-03-15)
Glyoxylate reductase/hydroxypyruvate reductase (GRHPR) is a key enzyme in the glyoxylate cycle. Its deficiency causes primary hyperoxaluria type 2. We first noticed that GRHPR was also lost in human hepatocellular carcinoma (HCC) and proliferative HCC cells. The aim of the
A novel mutation in the GRHPR gene in a Japanese patient with primary hyperoxaluria type 2.
Tatsuya Takayama et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 22(8), 2371-2374 (2007-05-19)
Veedamali S Subramanian et al.
American journal of physiology. Gastrointestinal and liver physiology, 304(12), G1079-G1086 (2013-04-20)
The human sodium-dependent vitamin C transporter 1 (hSVCT1) contributes to cellular uptake of ascorbic acid (AA). Although different aspects of hSVCT1 cell biology have been extensively studied, nothing is currently known about the broader hSVCT1 interactome that modulates its role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service