Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA021542

Sigma-Aldrich

Anti-TOP1MT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNA topoisomerase I, mitochondrial, Anti-TOP1mt

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$827.00

$827.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$827.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$827.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ARWEKEKHEDGVKWRQLEHKGPYFAPPYEPLPDGVRFFYEGRPVRLSVAAEEVATFYGRMLDHEYTTKEVFRKNFFNDWRKEMAVEEREVIKSLDKCDFTEIHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP1MT(116447)

General description

Mitochondrial topoisomerase 1 gene (TOP1MT) codes for type IB topoisomerase enzyme. TOP1mt gene consisting of 14 exons is mapped to human chromosome 8q24.3. Size of the protein varies between 60 and 70kDa. Optimum enzyme activity is at alkaline pH. As the name suggest this enzyme is exclusively targeted to mitochondria. TOP1MT enzyme is highly expressed in hematological cancer cells.

Immunogen

DNA topoisomerase I, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mitochondrial topoisomerase 1 (TOP1MT) enzyme relieves the topological stress of mitochondrial DNA by creating transient breaks in the DNA using a conserved tyrosine as the catalytic residue. Mutation in this gene might be responsible for mitochondrial disorders with neurodegenerative and myopathy components or to late-onset degenerative diseases such as Alzheimer′s disease, Parkinson′s syndrome, and diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85133

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Dalla Rosa et al.
The Journal of biological chemistry, 289(26), 18595-18602 (2014-05-07)
Mitochondrial topoisomerase I (Top1mt) is a type IB topoisomerase present in vertebrates and exclusively targeted to mitochondria. Top1mt relaxes mitochondrial DNA (mtDNA) supercoiling by introducing transient cleavage complexes wherein the broken DNA strand swivels around the intact strand. Top1mt cleavage
Screening of JAK2 V617F and MPL W515 K/L negative essential thrombocythaemia patients for mutations in SESN2, DNAJC17, ST13, TOP1MT, and NTRK1.
Carla Al Assaf et al.
British journal of haematology, 165(5), 734-737 (2014-03-13)
H Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(19), 10608-10613 (2001-08-30)
Tension generated in the circular mitochondrial genome during replication and transcription points to the need for mtDNA topoisomerase activity. Here we report a 601-aa polypeptide highly homologous to nuclear topoisomerase I. The N-terminal domain of this novel topoisomerase contains a
Hongliang Zhang et al.
Biochimie, 89(4), 474-481 (2006-12-13)
Mitochondria are the only organelles containing metabolically active DNA besides nuclei. By analogy with the nuclear topoisomerases, mitochondrial topoisomerase activities are probably critical for maintaining the topology of mitochondrial DNA during replication, transcription, and repair. Mitochondrial diseases include a wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service