Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA021483

Sigma-Aldrich

Anti-AHRR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Apoptosis-linked gene 2 protein, Anti-PDCD6, Anti-Probable calcium-binding protein ALG-2, Anti-Programmed cell death protein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

RQQVYISHLGHGVRGAQPHGRATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHRR(57491)

General description

Aryl hydrocarbon receptor repressor (AHRR) belongs to the basic helix-loop-helix period/aryl hydrocarbon receptor nuclear translocator/single-minded (bHLH-PAS) transcription factor family. AHRR gene of 60kb pairs long is localized to human chromosome 5p15.3 and contains 11 exons. AHRR protein shares structural similarity with AHR (aryl hydrocarbon receptor) but lacks a ligand-binding region (PAS B domain) and an activation domain.[1]

Immunogen

Programmed cell death protein 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aryl hydrocarbon receptor repressor (AHRR) functions as a negative feedback regulator of AHR (Aryl hydrocarbon receptor) pathway by binding with AHR nuclear translocator (ARNT) and XRE (xenobiotic response element), which have competing binding sites for AHR (Aryl hydrocarbon receptor). Therefore, it is known as a transcriptional repressor of AHR. AHRR also plays a vital role in human reproductive system and regulation of cell growth. Consequently, mutations in this gene cause infertility in men and endometriosis in women. It may also functions as a tumor suppressor gene in various types of human cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74245.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Haarmann-Stemmann et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(12), 2262-2269 (2007-09-25)
The aryl hydrocarbon receptor repressor (AhRR) is a member of the aryl hydrocarbon receptor (AhR) signaling cascade, which mediates dioxin toxicity and is involved in regulation of cell growth and differentiation. The AhRR was described as a feedback modulator, which
Sibel I Karchner et al.
Molecular and cellular biology, 29(13), 3465-3477 (2009-04-22)
The aryl hydrocarbon receptor (AHR) repressor (AHRR) inhibits AHR-mediated transcription and has been associated with reproductive dysfunction and tumorigenesis in humans. Previous studies have characterized the repressor function of AHRRs from mice and fish, but the human AHRR ortholog (AHRR(715))
T Baba et al.
The Journal of biological chemistry, 276(35), 33101-33110 (2001-06-26)
The aryl hydrocarbon receptor (AhR) repressor (AhRR) gene has been isolated and characterized from a mouse genomic library. The gene is distributed as 11 exons in a total length of about 60 kilobase pairs. Fluorescence in situ hybridization analysis has
Laura MacPherson et al.
International journal of molecular sciences, 15(5), 7939-7957 (2014-05-09)
The aryl hydrocarbon receptor (AHR) regulates the toxic effects of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). The AHR repressor (AHRR) is an AHR target gene and functions as a ligand-induced repressor of AHR; however, its mechanism of inhibition is controversial. Recently, we reported that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service