Skip to Content
MilliporeSigma
All Photos(2)

Documents

HPA021410

Sigma-Aldrich

Anti-PHF12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PHD factor 1, Anti-PHD finger protein 12, Anti-Pf1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GGAVNMCYRTLYIGTGADMDVCLTNYGHCNYVSGKHACIFYDENTKHYELLNYSEHGTTVDNVLYSCDFSEKTPPTPPSSIVAKVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF12(57649)

General description

The gene PHF12 (PHD finger protein 12) is mapped to human chromosome 17q11.2. The protein contains two plant homeodomain (PHD) zinc fingers. It is expressed in heart, brain, lung, liver and testis. PHF12 localizes in the nucleus. PHF12 is commonly referred to as PF1 (PHD factor 1).

Immunogen

PHD finger protein 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PHF12 (PHD finger protein 12) is involved in transcriptional regulation. It interacts with Sin3A (SIN3 transcription regulator family member A) complex to repress transcription. Similarly, PHF12-mediated recruitment of TLE (transducin-like enhancer) complex represses transcription.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75593

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Yochum et al.
Molecular and cellular biology, 21(13), 4110-4118 (2001-06-08)
The mSin3A-histone deacetylase corepressor is a multiprotein complex that is recruited by DNA binding transcriptional repressors. Sin3 has four paired amphipathic alpha helices (PAH1 to -4) that are protein-protein interaction motifs and is the scaffold upon which the complex assembles.
Fei Chen et al.
Investigative ophthalmology & visual science, 56(1), 544-551 (2014-12-20)
Intraocular pressure (IOP) is an important clinical parameter in the evaluation of ocular health. Elevated IOP is a major risk factor for primary open-angle glaucoma (POAG). The goal of this study was to identify rare and less common variants that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service