Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

HPA021230

Sigma-Aldrich

Anti-GOLGA2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cis-Golgi matrix protein GM130, Anti-Gm130 autoantigen, Anti-Golgin subfamily A member 2, Anti-Golgin-95

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LQQYVAAYQQLTSEKEVLHNQLLLQTQLVDQLQQQEAQGKAVAEMARQELQETQERLEAATQQNQQLRAQLSLMAHPG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOLGA2(2801)

General description

The gene GOLGA2 (golgin subfamily A member 2) is mapped to human chromosome 9q34.11. It belongs to the family of coiled-coil golgins. The protein cycles between the cis-Golgi compartments and the ER (endoplasmic reticulum)-to-Golgi carriers.

Immunogen

Golgin subfamily A member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Golgi apparatus ribbon formation requires specific membrane-fusion events. GOLGA2 (golgin subfamily A member 2) is crucial for lateral cisternal-fusion reactions and helps in proper distribution of enzymes in the Golgi apparatus. It also participates in endoplasmic reticulum-to-Golgi transport and mitotic Golgi apparatus fragmentation. It interacts with and stabilizes GRASP65 (golgi peripheral membrane protein p65).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75375

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierfrancesco Marra et al.
Molecular biology of the cell, 18(5), 1595-1608 (2007-02-23)
The Golgi complex in mammalian cells forms a continuous ribbon of interconnected stacks of flat cisternae. We show here that this distinctive architecture reflects and requires the continuous input of membranes from the endoplasmic reticulum (ER), in the form of
Nobuhiro Nakamura
Journal of pharmacological sciences, 112(3), 255-264 (2010-03-04)
GM130 is a peripheral membrane protein strongly attached to the Golgi membrane and is isolated from the detergent and salt resistant Golgi matrix. GM130 is rich in coiled-coil structures and predicted to take a rod-like shape. Together with p115, giantin
Sabrina Rivero et al.
The EMBO journal, 28(8), 1016-1028 (2009-02-27)
We report that microtubule (MT) nucleation at the Golgi apparatus requires AKAP450, a centrosomal gamma-TuRC-interacting protein that also forms a distinct network associated with the Golgi. Depletion of AKAP450 abolished MT nucleation at the Golgi, whereas depletion of the cis-Golgi
Nozomu Yanaihara et al.
Cancer cell, 9(3), 189-198 (2006-03-15)
MicroRNA (miRNA) expression profiles for lung cancers were examined to investigate miRNA's involvement in lung carcinogenesis. miRNA microarray analysis identified statistical unique profiles, which could discriminate lung cancers from noncancerous lung tissues as well as molecular signatures that differ in
Manojkumar A Puthenveedu et al.
Nature cell biology, 8(3), 238-248 (2006-02-21)
The mammalian Golgi apparatus exists as stacks of cisternae that are laterally linked to form a continuous membrane ribbon, but neither the molecular requirements for, nor the purpose of, Golgi ribbon formation are known. Here, we demonstrate that ribbon formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service