Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA019708

Sigma-Aldrich

Anti-SMU1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Smu-1 suppressor of mec-8 and unc-52 protein homolog, Anti-WD40 repeat-containing protein SMU1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

SIEIESSDVIRLIMQYLKENSLHRALATLQEETTVSLNTVDSIESFVADINSGHWDTVLQAIQSLKLPDKTLIDLYEQVVLELIELRELGAARSLLRQTDPMIML

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMU1(55234)

General description

SMU1 (Smu-1 suppressor of mec-8 and unc-52 homolog, C. elegans) is an evolutionarily conserved gene belonging to the WD40-repeat protein family.

Immunogen

WD40 repeat-containing protein SMU1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SMU1 (Smu-1 suppressor of mec-8 and unc-52 homolog, C. elegans) participates in pre-mRNAs splicing as a splicing factor. SMU1, in conjugation with splicing factor RED, regulates gene expression of influenza virus. It is also involved in the positive regulation of splicing of NS1 (Nonstructural protein 1) protein responsible for viral multiplication. SMU1 plays a vital role in the maintenance of genomic structural integrity by acting as a chromatin-binding protein. It also negatively controls DNA synthesis pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75231

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Fournier et al.
PLoS pathogens, 10(6), e1004164-e1004164 (2014-06-20)
Influenza A viruses are major pathogens in humans and in animals, whose genome consists of eight single-stranded RNA segments of negative polarity. Viral mRNAs are synthesized by the viral RNA-dependent RNA polymerase in the nucleus of infected cells, in close
Laifeng Ren et al.
Biochemical and biophysical research communications, 436(2), 192-198 (2013-06-04)
Smu1 is an evolutionarily conserved gene that encodes a member of the WD40-repeat protein family. Disruption of Smu1 function leads to multiple cellular defects including chromosomal instability, aberrant DNA replication and alternative RNA splicing events. In this paper, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service