Skip to Content
MilliporeSigma
All Photos(6)

Documents

HPA019674

Sigma-Aldrich

Anti-NPAS2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MOP4, Anti-PASD4, Anti-bHLHe9, Anti-neuronal PAS domain protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SKPEFIVCTHSVVSYADVRVERRQELALEDPPSEALHSSALKDKGSSLEPRQHFNTLDVGASGLNTSHSPSASSRSSHKSSHTAMSEPTSTPTKLMAEASTPALPRSATLPQELPVP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPAS2(4862)

General description

NPAS2 (Neuronal PAS domain protein 2) is a largest circadian gene encoded helix-loop-helix-PAS class of transcription factor. It is located on chromosome 2q11.2. It is mostly expressed in the mammalian forebrain.

Immunogen

neuronal PAS domain protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NPAS2 (Neuronal PAS domain protein 2) is a transcription factor highly involved in the regulation of circadian rhythm. Its PAS domains directly bind to the heme as a prosthetic group to control DNA binding in vitro. In the maintenance of circadian behavioral rhythms, NPAS2 heterodimerize with BMAL1, which stimulates transcriptional expression of the circadian genes required for biological rhythms in many organisms.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84800

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elhadji M Dioum et al.
Science (New York, N.Y.), 298(5602), 2385-2387 (2002-11-26)
Neuronal PAS domain protein 2 (NPAS2) is a mammalian transcription factor that binds DNA as an obligate dimeric partner of BMAL1 and is implicated in the regulation of circadian rhythm. Here we show that both PAS domains of NPAS2 bind
Yong Zhu et al.
Breast cancer research and treatment, 107(3), 421-425 (2007-04-25)
Three known non-synonymous polymorphisms (Ala394Thr, Ser471Leu and Pro690Ala) in the largest circadian gene, Neuronal PAS domain protein 2 (NPAS2), were genotyped in a breast cancer case-control study conducted in Connecticut, USA (431 cases and 476 controls). We found that women
Jason P DeBruyne et al.
Nature neuroscience, 10(5), 543-545 (2007-04-10)
Heterodimers of CLOCK and BMAL1, bHLH-PAS transcription factors, are believed to be the major transcriptional regulators of the circadian clock mechanism in mammals. However, a recent study shows that CLOCK-deficient mice continue to exhibit robust behavioral and molecular rhythms. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service