Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA019669

Sigma-Aldrich

Anti-RABEP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Rab GTPase-binding effector protein 1, Anti-Rabaptin-4, Anti-Rabaptin-5, Anti-Rabaptin-5alpha, Anti-Renal carcinoma antigen NY-REN-17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SREQVSEELVRLQKDNDSLQGKHSLHVSLQQAEDFILPDTTEALRELVLKYREDIINVRTAADHVEEKLKAEILFLKEQIQAEQCLKENLEETLQL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RABEP1(9135)

General description

RABEP1 (Rabaptin, RAB GTPase binding effector protein 1) is a novel 100kDa cytosolic protein consisting of coiled-coil domain. It is colocalized on large early endosomes.

Immunogen

Rab GTPase-binding effector protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RABEP1 (Rabaptin, RAB GTPase binding effector protein 1) is a Rab effector protein. During membrane docking and fusion, its C-terminal domain form a dyad-symmetric Rab5-Rabaptin5(2)-Rab5 ternary complex by binding to their switch and interswitch regions. Any mutation in Rab5-Rabaptin5(2)-Rab5 binding causes enlarged endosomes. RABEP1 also participates in K(V)10.1 potassium channel trafficking on the cell surface. It has been reported that silencing of RABEP1 shows induced down-regulation of recycling of K(V)10.1 channel.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74234

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangyu Zhu et al.
Nature structural & molecular biology, 11(10), 975-983 (2004-09-21)
Rab5 is a small GTPase that regulates early endosome fusion. We present here the crystal structure of the Rab5 GTPase domain in complex with a GTP analog and the C-terminal domain of effector Rabaptin5. The proteins form a dyad-symmetric Rab5-Rabaptin5(2)-Rab5
H Stenmark et al.
Cell, 83(3), 423-432 (1995-11-03)
We have identified a novel 100 kDa coiled-coil protein, rabaptin-5, that specifically interacts with the GTP form of the small GTPase Rab5, a potent regulator of endocytic transport. It is mainly cytosolic, but a fraction colocalizes with Rab5 to early
Milena Ninkovic et al.
FEBS letters, 586(19), 3077-3084 (2012-07-31)
K(V)10.1 is a potassium channel expressed in brain and implicated in tumor progression. We have searched for proteins interacting with K(V)10.1 and identified Rabaptin-5, an effector of the Rab5 GTPase. Both proteins co-localize on large early endosomes induced by Rab5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service