Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA019194

Sigma-Aldrich

Anti-SCAMP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Secretory carrier membrane protein 2, Anti-Secretory carrier-associated membrane protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QRVHSLYRRTGASFQQAQEEFSQGIFSSRTFHRAASSAAQGAFQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCAMP2(10066)

General description

The gene SCAMP2 (secretory carrier-associated membrane protein 2) is mapped to human chromsome15q24. It belongs to SCAMP gene family.

Immunogen

Secretory carrier-associated membrane protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SCAMP2 (secretory carrier-associated membrane protein 2) interacts with (Na+,K+)/H+ exchanger (NHE7) and is responsible for transfer of NHE7 between recycling vesicles and the trans-Golgi network. It also interacts with dopamine and serotonin transporters, and regulates their cell surface expression. SCAMP2 interaction with brain-enriched Na+/H+ exchanger, NHE5, allows surface targeting of NHE5 in an ARF6 (ADP-ribosylation factor 6)-dependent manner.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73122

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulo J C Lin et al.
Journal of cell science, 118(Pt 9), 1885-1897 (2005-04-21)
The mammalian (Na(+),K(+))/H(+) exchanger NHE7 resides chiefly in the trans-Golgi network (TGN) and post-Golgi vesicles where it is thought to contribute to organellar pH homeostasis. However, the mechanisms that underlie the targeting and regulation of NHE7 are unknown. To gain
Graham H Diering et al.
The Journal of biological chemistry, 284(20), 13892-13903 (2009-03-12)
NHE5 is a brain-enriched Na(+)/H(+) exchanger that dynamically shuttles between the plasma membrane and recycling endosomes, serving as a mechanism that acutely controls the local pH environment. In the current study we show that secretory carrier membrane proteins (SCAMPs), a
Susanne Haemmerling et al.
British journal of haematology, 157(2), 180-187 (2012-02-03)
Transient myeloproliferative disorder (TMD) of the newborn and acute megakaryoblastic leukaemia (AMKL) in children with Down syndrome (DS) represent paradigmatic models of leukaemogenesis. Chromosome 21 gene dosage effects and truncating mutations of the X-chromosomal transcription factor GATA1 synergize to trigger
Anja W Fjorback et al.
Biochemical and biophysical research communications, 406(2), 165-170 (2011-02-08)
The monoamine transporters for dopamine (DAT), norepinephrine (NET) and serotonin (SERT) facilitate the homeostatic balance of neurotransmitters in the synaptic cleft and thus, play a fundamental role in regulating neuronal activity. Despite the importance of these monoamine transporters in controlling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service