Skip to Content
MilliporeSigma
All Photos(1)

Documents

HPA018525

Sigma-Aldrich

Anti-BNC2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein basonuclin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

YENESESSEPKLGEESMEGDEHIHSEVSEKVLMNSERPDENHSEPSHQDVIKVKEEFTDPTYDMFYMSQYGLYNGGGASMAALHESFTSSLNYGSPQKFSPEGDLCSSPDPKICYVCKKSFKSSYSVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BNC2(54796)

General description

The gene basonuclin-2 (BNC2) is mapped to human chromosome 9p22. It belongs to basonuclin zinc-finger family of transcription factors. BNC2 transcripts are mainly present in tissues of the reproductive system (ovary and testis), kidney and skin. The protein is localized in the nucleus.

Immunogen

Zinc finger protein basonuclin-2 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Basonuclin-2 (BNC2) is associated with skin color variation and skin cancer risk. It also contributes to the occurrence of facial pigmented spots during aging. BNC2 has been shown to be associated with epithelial ovarian cancer. Knockdown of BNC2 causes distal urethral defects in mouse model.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73749

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey J Winham et al.
Genetic epidemiology, 38(5), 457-466 (2014-05-24)
Due to its potential as a biomarker for early cancer detection, blood-based DNA methylation (DNAm) is of interest in cancer research. Specifically, highly predictive mechanisms for early detection of epithelial ovarian cancer (EOC) are desired, so previous studies have compared
Leonie C Jacobs et al.
The Journal of investigative dermatology, 135(7), 1735-1742 (2015-02-24)
Facial pigmented spots are a common skin aging feature, but genetic predisposition has yet to be thoroughly investigated. We conducted a genome-wide association study for pigmented spots in 2,844 Dutch Europeans from the Rotterdam Study (mean age: 66.9±8.0 years; 47%
Rose-Anne Romano et al.
Genomics, 83(5), 821-833 (2004-04-15)
We used a bioinformatics approach to identify Basonuclin2, the second member of the Basonuclin zinc-finger family of transcription factors. The mouse Basonuclin2 protein consists of 1049 amino acids and contains three pairs of zinc fingers in the C-terminus that show
Melissa A Buckley et al.
Cancer research, 79(3), 467-481 (2018-11-30)
Genome-wide association studies have identified 40 ovarian cancer risk loci. However, the mechanisms underlying these associations remain elusive. In this study, we conducted a two-pronged approach to identify candidate causal SNPs and assess underlying biological mechanisms at chromosome 9p22.2, the
Leonie C Jacobs et al.
Human genetics, 132(2), 147-158 (2012-10-12)
Natural variation in human skin pigmentation is primarily due to genetic causes rooted in recent evolutionary history. Genetic variants associated with human skin pigmentation confer risk of skin cancer and may provide useful information in forensic investigations. Almost all previous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service