Skip to Content
MilliporeSigma
All Photos(9)

Documents

HPA018520

Sigma-Aldrich

Anti-CCT8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CCT-theta, Anti-Renal carcinoma antigen NY-REN-15, Anti-T-complex protein 1 subunit theta, Anti-TCP-1-theta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LPNLVCCSAKNLRDIDEVSSLLRTSIMSKQYGNEVFLAKLIAQACVSIFPDSGHFNVDNIRVCKILGSGISSSSVLHGMVFKKETEGDVT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCT8(10694)

General description

The gene T-complex protein 1 subunit θ (CCT8) is mapped to human chromosome 21q22.11. It belongs to CCT family of chaperonins. The protein is localized in the cytoplasm.

Immunogen

T-complex protein 1 subunit theta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CCT proteins are crucial for protein folding in ATP-dependent manner. T-complex protein 1 subunit θ (CCT8) is part of a complex including chaperonin-like BBS (Bardet-Biedl syndrome phenotype) proteins (BBS6, BBS10, and BBS12) that mediate BBSome assembly, required for transporting vesicles to the cilia. Knockdown of CCT proteins in zebrafish results in BBS phenotype. CCT8 has been suggested as an oncogene. It is up-regulated in hepatocellular carcinoma and positively correlates with tumor size. Depletion of CCT8 inhibits cell proliferation and block S-phase entry.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73929

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumori Kawakami et al.
Oncology reports, 16(3), 521-531 (2006-07-26)
Large-scale gene expression profiling is an effective strategy for understanding the progression of bladder cancer (BC). The aim of this study was to identify genes that are expressed differently in the course of BC progression and to establish new biomarkers
Xiaodong Huang et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 122(11), 1070-1079 (2014-05-28)
The development of molecular pathogenesis of hepatocellular carcinoma (HCC) is complex and involves alterations in the expression and conformation of assorted oncoproteins and tumor suppressors. Chaperonin containing TCP1 (CCT) is a cytolic molecular chaperone complex that is required for the
Seongjin Seo et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(4), 1488-1493 (2010-01-19)
Bardet-Biedl syndrome (BBS) is a human genetic disorder resulting in obesity, retinal degeneration, polydactyly, and nephropathy. Recent studies indicate that trafficking defects to the ciliary membrane are involved in this syndrome. Here, we show that a novel complex composed of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service