Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA018135

Sigma-Aldrich

Anti-KIAA1671 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CTA-221G9.5, Anti-Uncharacterized protein KIAA1671, Anti-Z99916.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PISHSLRRSRFSESESRSPLEDETDNTWMFKDSTEEKSPRKEESDEEETASKAERTPVSHPQRMPAFPGMDPAVLKAQLHKRPEVDSPGETPSWAPQPKSPKSPFQPGVLGSRVLPSSMDKDERSDEPSPQWLKELKSKKRQSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene KIAA1671 has been mapped to human chromosome band 22q11.23.

Immunogen

Uncharacterized protein KIAA1671 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene KIAA1671 has been shown to participate in multiple carcinomas. KIAA1671 is down-regulated in classical and follicular variants of papillary thyroid carcinoma. The protein is also identified as breast cancer-associated auto-antigen. It is shown to distinguish ductal carcinoma in situ from invasive ductal carcinoma of the breast and is suggested to be a potential biomarker for the diagnosis of breast cancer. The gene is up-regulated in metachromatic leukodystrophy pateints, a lysosomal storage disorder. Over expression of miR-137 (one of the leading candidate schizophrenia susceptibility gene) in human neural progenitor cell line CTXOE03, down-regulates the predicted target gene KIAA1671.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73413

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Cesani et al.
Annals of neurology, 75(1), 127-137 (2013-11-19)
To facilitate development of novel disease-modifying therapies for lysosomal storage disorder (LSDs) characterized by nervous system involvement such as metachromatic leukodystrophy (MLD), molecular markers for monitoring disease progression and therapeutic response are needed. To this end, we sought to identify
Félix Fernández-Madrid et al.
Cancer research, 64(15), 5089-5096 (2004-08-04)
We report on the identification of autoantigens commonly recognized by sera from patients with breast cancer. We selected ten sera from patients with invasive ductal carcinoma (IDC) of the breast with high titer IgG autoantibodies for biopanning of a T7
Félix Fernández Madrid
Cancer letters, 230(2), 187-198 (2005-11-22)
A plethora of promising breast cancer-associated autoantigens have been cloned by immunoscreening cDNA expression libraries with breast cancer sera or identified using proteomics, yet no biomarkers, whether individual autoantigens or panels of antigens developed using antibody-based methods have been validated
Matthew J Hill et al.
Schizophrenia research, 153(1-3), 225-230 (2014-02-22)
MIR137, transcribed as the microRNA miR-137, is one of the leading candidate schizophrenia susceptibility genes to arise from large genome-wide association studies (GWAS) of the disorder. Recent data suggest that miR-137 modulates the expression of other schizophrenia susceptibility genes. Although
Yusuf Ziya Igci et al.
Endocrine pathology, 22(2), 86-96 (2011-04-22)
Fine-needle aspiration biopsy (FNA) is currently the best initial diagnostic test for evaluation of a thyroid nodule. FNA cytology cannot discriminate between benign and malignant thyroid nodules in up to 30% of thyroid nodules. Therefore, an adjunct to FNA is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service