Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA017864

Sigma-Aldrich

Anti-SERPINA6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CBG, Anti-Corticosteroid-binding globulin precursor, Anti-Serpin A6, Anti-Transcortin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

TVFFILPDKGKMNTVIAALSRDTINRWSAGLTSSQVDLYIPKVTISGVYDLGDVLEEMGIADLFTNQANFSRITQDAQLKSSKVVHKAVLQLNEEGVDTAGSTGVTLNLTSKPIILRFNQPFIIMIFDHFTWSSLFLARVMN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINA6(866)

General description

Serine protease inhibitor A6 (Serpin A6) is a non-inhibitory protein belonging to the serpin superfamily of serine protease inhibitors. It is a glycosylated plasma protein consisting of 383 amino acids. SERPINA6 has six sites for N-glycosylation and the gene encoding it is localized on chromosome 14q32.1. It is also referred as CBG (Corticosteroid-binding globulin).

Immunogen

Corticosteroid-binding globulin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Serine protease inhibitor A6 (Serpin A6) carries most of the cortisol in circulation. Neutrophil elastase can cleave Serpin A6 at its exposed reactive centre loop. This decreases its affinity for cortisol, thus aiding in the release of cortisol at the site of inflammation. SERPINA6 is an important plasma binding protein of glucocorticoids and transports them in the blood thus controlling the tissue availability of these hormones.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70433

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John G Lewis et al.
The Journal of steroid biochemistry and molecular biology, 149, 53-57 (2015-02-01)
Corticosteroid-binding globulin (CBG) is a non-inhibitory member of the serpin superfamily of serine protease inhibitors and carries the majority of cortisol in circulation. It can be cleaved by neutrophil elastase at its exposed reactive centre loop which decreases its affinity
P Rollini et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 8(12), 913-916 (1997-12-13)
The genes encoding alpha 1-antitrypsin (alpha 1AT; gene symbol PI) and corticosteroid-binding globulin (CBG) are part of a cluster of structurally related serine protease inhibitor (serpin) genes on human Chromosome (Chr) 14q32.1. This cluster also includes the genes encoding alpha
Bernd R Gardill et al.
PloS one, 7(12), e52759-e52759 (2013-01-10)
Corticosteroid-binding globulin (CBG) transports glucocorticoids and progesterone in the blood and thereby modulates the tissue availability of these hormones. As a member of the serine protease inhibitor (SERPIN) family, CBG displays a reactive center loop (RCL) that is targeted by
Marc Simard et al.
The Journal of clinical endocrinology and metabolism, 100(1), E129-E139 (2014-10-17)
Corticosteroid-binding globulin (CBG) is encoded by SERPINA6. It is the major plasma binding protein of glucocorticoids and regulates plasma cortisol levels and bioavailability in humans. Several proteases target CBG and disrupt its steroid-binding properties. To date, most genetic deficiencies that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service